The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020046.3 | NP_064430.1 | 4 - 5 | 819 | 89 |
GenBank mRNA | AF029667.2 | - | 4 - 5 | 524 | 89 |
AK030798.1 | - | - | 403 | 89 | |
BC019542.1 | - | 4 - 5 | 529 | 89 | |
BC027829.1 | - | 4 - 5 | 780 | 89 | |
BC045206.1 | - | 4 - 5 | 769 | 89 |