Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr12
Entrez Gene ID
57750
Gene Name
WD repeat domain 12
Gene Aliases
4933402C23Rik, Ytm1, Ytm1p
Chromosome Location
Chr.1: 60076868 - 60098500 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199060.1 NP_001185989.1 7 - 8 1151 101
NM_001199061.1 NP_001185990.1 7 - 8 883 101
NM_021312.5 NP_067287.1 6 - 7 973 101
GenBank mRNA AB041608.1 - 7 - 8 1152 101
AF239765.1 - 6 - 7 629 101
AK007500.1 - 6 - 7 750 101
AK012022.1 - 6 - 7 727 101
AK016611.1 - 7 - 8 827 101
AK031436.1 - 6 - 7 736 101
AK149085.1 - 6 - 7 655 101
AK161299.1 - 7 - 8 896 101
AY059431.1 - 6 - 7 664 101
BC004748.1 - 6 - 7 807 101
BC052386.1 - 6 - 7 944 101
BC096651.1 - 6 - 7 718 101

View Full Product Details