Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Wdr12
Entrez Gene ID
57750
Gene Name
WD repeat domain 12
Gene Aliases
4933402C23Rik, Ytm1, Ytm1p
Chromosome Location
Chr.1: 60076868 - 60098500 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001199060.1 NP_001185989.1 13 - 14 1728 82
NM_001199061.1 NP_001185990.1 13 - 14 1460 82
NM_021312.5 NP_067287.1 12 - 13 1550 82
GenBank mRNA AB041608.1 - 13 - 14 1729 82
AF239765.1 - 12 - 13 1206 82
AK007500.1 - 12 - 13 1327 82
AK016611.1 - 13 - 14 1404 82
AK149085.1 - 12 - 13 1232 82
AK161299.1 - 13 - 14 1473 82
AY059431.1 - 12 - 13 1241 82
AY059432.1 - 5 - 6 546 82
BC004748.1 - 12 - 13 1384 82
BC052386.1 - 12 - 13 1521 82
BC096651.1 - 12 - 13 1295 82

View Full Product Details