Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdc25b
Entrez Gene ID
12531
Gene Name
cell division cycle 25B
Gene Aliases
AI604853
Chromosome Location
Chr.2: 131186436 - 131198511 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111075.4 NP_001104545.1 2 - 3 557 94
NM_023117.4 NP_075606.1 2 - 3 557 94
GenBank mRNA AK004836.1 - 2 - 3 514 94
AK043730.1 - 2 - 3 548 94
AK153903.1 - 2 - 3 558 94
BC048164.1 - 2 - 3 554 94
BC057568.1 - 2 - 3 428 94
BQ561430.1 - 2 - 3 213 94
BU057369.1 - 2 - 3 554 94
S93521.1 - 2 - 3 426 94

View Full Product Details