The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139511.1 | NP_001132983.1 | 4 - 5 | 935 | 72 |
NM_001139512.1 | NP_001132984.1 | 5 - 6 | 701 | 72 | |
NM_001139513.1 | NP_001132985.1 | 5 - 6 | 983 | 72 | |
NM_023130.3 | NP_075619.2 | 5 - 6 | 1018 | 72 | |
GenBank mRNA | AK014356.1 | - | 5 - 6 | 771 | 72 |
AK135386.1 | - | 4 - 5 | 694 | 72 | |
AK154787.1 | - | 4 - 5 | 692 | 72 | |
AK168781.1 | - | 5 - 6 | 775 | 72 | |
BC004851.1 | - | 4 - 5 | 679 | 72 | |
BC016587.1 | - | 5 - 6 | 767 | 72 | |
L17076.1 | - | 5 - 6 | 748 | 72 | |
S72641.1 | - | 5 - 6 | 969 | 72 |