The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029573.2 | NP_083849.1 | 10 - 11 | 1017 | 65 |
GenBank mRNA | AK003393.1 | - | 7 - 8 | 910 | 65 |
AK010065.1 | - | 10 - 11 | 1041 | 65 | |
AK032787.1 | - | 10 - 11 | 1041 | 65 | |
AK047951.1 | - | 10 - 11 | 1039 | 65 | |
AK117103.1 | - | 10 - 11 | 1053 | 65 | |
AK146717.1 | - | 7 - 8 | 881 | 65 | |
AK150618.1 | - | 10 - 11 | 1048 | 65 | |
AK151304.1 | - | 10 - 11 | 1042 | 65 | |
AK152353.1 | - | 10 - 11 | 1051 | 65 | |
AK153459.1 | - | 10 - 11 | 1042 | 65 | |
AK158646.1 | - | 10 - 11 | 1195 | 65 | |
AK159051.1 | - | 10 - 11 | 1114 | 65 | |
AK159666.1 | - | 10 - 11 | 1046 | 65 | |
AK160054.1 | - | 10 - 11 | 1046 | 65 | |
AK164038.1 | - | 11 - 12 | 1144 | 65 | |
AK167822.1 | - | 10 - 11 | 1047 | 65 | |
AK168034.1 | - | 10 - 11 | 1037 | 65 | |
AK168049.1 | - | 10 - 11 | 1042 | 65 | |
AK168149.1 | - | 10 - 11 | 1048 | 65 | |
AK169152.1 | - | 10 - 11 | 1015 | 65 | |
AK196914.1 | - | 1 - 2 | 131 | 65 | |
BC034273.1 | - | 10 - 11 | 1007 | 65 | |
BC049956.1 | - | 10 - 11 | 1017 | 65 |