Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dok5
Entrez Gene ID
76829
Gene Name
docking protein 5
Gene Aliases
2700055C10Rik
Chromosome Location
Chr.2: 170731807 - 170879775 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163686.1 NP_001157158.1 1 - 2 381 105
NM_029761.4 NP_084037.3 1 - 2 381 105
GenBank mRNA AF418208.1 - 1 - 2 243 105
BC066822.1 - 1 - 2 381 105
BY248468.1 - 1 - 2 297 105

View Full Product Details