Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Setd7
Entrez Gene ID
73251
Gene Name
SET domain containing (lysine methyltransferase) 7
Gene Aliases
1600028F23Rik, H3K4MT, KMT7, Set7, Set7/9, mKIAA1717
Chromosome Location
Chr.3: 51515318 - 51560823 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080793.5 NP_542983.3 3 - 4 636 101
GenBank mRNA AF448509.1 - 3 - 4 418 101
AK048924.1 - 3 - 4 473 101
AK129428.1 - 3 - 4 425 101
AK147316.1 - 3 - 4 692 101
AK147413.1 - 3 - 4 462 101
AK147422.1 - 3 - 4 637 101
AK147667.1 - 3 - 4 671 101
AK170161.1 - 3 - 4 480 101
BC050190.1 - 3 - 4 460 101

View Full Product Details