The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008452.2 | NP_032478.2 | 1 - 2 | 91 | 75 |
GenBank mRNA | BC138737.1 | - | 1 - 2 | 99 | 75 |
BC138738.1 | - | 1 - 2 | 99 | 75 | |
BF023555.1 | - | 1 - 2 | 73 | 75 | |
U25096.1 | - | 1 - 2 | 120 | 75 |