The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008593.3 | NP_032619.1 | 1 - 1 | 170 | 65 |
GenBank mRNA | AF023915.1 | - | 1 - 1 | 148 | 65 |