The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008979.2 | NP_033005.1 | 20 - 21 | 2469 | 90 |
NR_104070.1 | 22 - 23 | 2486 | 90 | ||
GenBank mRNA | AI327325.1 | - | 1 - 2 | 90 | |
AK153812.1 | - | 22 - 23 | 2487 | 90 | |
BC055377.1 | - | 20 - 21 | 2391 | 90 | |
M90388.1 | - | 20 - 21 | 2454 | 90 |