The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001104616.1 | NP_001098086.1 | 4 - 5 | 345 | 138 |
NM_001104617.1 | NP_001098087.1 | 4 - 5 | 751 | 138 | |
NM_009041.3 | NP_033067.2 | 4 - 5 | 751 | 138 | |
GenBank mRNA | AK016711.1 | - | 4 - 5 | 422 | 138 |
AK088292.1 | - | 4 - 5 | 458 | 138 | |
AK146423.1 | - | 4 - 5 | 409 | 138 | |
AK156370.1 | - | 4 - 5 | 437 | 138 | |
AK162204.1 | - | 4 - 5 | 323 | 138 | |
AK168449.1 | - | 4 - 5 | 422 | 138 | |
BC002041.1 | - | 4 - 5 | 396 | 138 | |
BC026786.1 | - | 4 - 5 | 424 | 138 | |
BC034438.1 | - | 4 - 5 | 577 | 138 | |
BC053417.1 | - | 4 - 5 | 425 | 138 | |
DV662898.1 | - | 4 - 5 | 345 | 138 | |
X60672.1 | - | 4 - 5 | 364 | 138 |