The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009045.4 | NP_033071.1 | 8 - 9 | 1224 | 67 |
GenBank mRNA | AK089489.1 | - | 8 - 9 | 969 | 67 |
AK144565.1 | - | 8 - 9 | 969 | 67 | |
AK154630.1 | - | 8 - 9 | 969 | 67 | |
AK155321.1 | - | 8 - 9 | 1233 | 67 | |
AK155889.1 | - | 8 - 9 | 966 | 67 | |
AK157076.1 | - | 8 - 9 | 956 | 67 | |
AK158072.1 | - | 8 - 9 | 1234 | 67 | |
BC003818.1 | - | 8 - 9 | 983 | 67 | |
BC094053.1 | - | 8 - 9 | 1224 | 67 | |
DQ020177.1 | - | 6 - 7 | 793 | 67 | |
M61909.1 | - | 8 - 9 | 939 | 67 |