Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
St3gal4
Entrez Gene ID
20443
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 4
Gene Aliases
Siat4c
Chromosome Location
Chr.9: 35046576 - 35117445 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009178.3 NP_033204.2 5 - 6 448 59
GenBank mRNA AB061305.1 - 5 - 6 396 59
BC011121.1 - 5 - 6 393 59
BC050773.1 - 5 - 6 448 59
D28941.1 - 5 - 6 403 59
X95809.1 - 4 - 5 275 59

View Full Product Details