The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010957.4 | NP_035087.3 | 1 - 2 | 387 | 106 |
GenBank mRNA | AF000669.1 | - | 1 - 2 | 265 | 106 |
AF003596.1 | - | 1 - 2 | 352 | 106 | |
AK146870.1 | - | 1 - 2 | 387 | 106 | |
AK165875.1 | - | 1 - 2 | 339 | 106 | |
AK167456.1 | - | 1 - 2 | 364 | 106 | |
BC138733.1 | - | 1 - 2 | 343 | 106 | |
BC138736.1 | - | 1 - 2 | 343 | 106 | |
U88621.1 | - | 1 - 2 | 355 | 106 | |
U96711.1 | - | 1 - 2 | 352 | 106 | |
Y11247.1 | - | 1 - 2 | 318 | 106 | |
Y13479.1 | - | 1 - 2 | 388 | 106 |