The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011825.1 | NP_035955.1 | 1 - 2 | 240 | 57 |
GenBank mRNA | AB011030.1 | - | 1 - 2 | 240 | 57 |
AK142008.1 | - | 1 - 2 | 242 | 57 | |
AK161827.1 | - | 1 - 2 | 220 | 57 | |
BC079905.1 | - | 1 - 2 | 221 | 57 |