The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018867.5 | NP_061355.3 | 2 - 3 | 554 | 80 |
GenBank mRNA | AF017639.1 | - | 2 - 3 | 555 | 80 |
AK019509.1 | - | 2 - 3 | 553 | 80 | |
AK038874.1 | - | 2 - 3 | 437 | 80 | |
BC021444.1 | - | 2 - 3 | 476 | 80 |