Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Amotl2
Entrez Gene ID
56332
Gene Name
angiomotin-like 2
Gene Aliases
AW549739, C79691, Lccp, MASCOT, mKIAA0989
Chromosome Location
Chr.9: 102717294 - 102733418 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019764.2 NP_062738.2 7 - 8 2033 62
GenBank mRNA AF175967.1 - 7 - 8 1316 62
AF175968.1 - 5 - 6 2173 62
AK040912.1 - 7 - 8 2114 62
AK077535.1 - 7 - 8 2029 62
AK141348.1 - 7 - 8 2033 62
AK164279.1 - 7 - 8 2102 62
AK164819.1 - 7 - 8 2033 62
AK169419.1 - 7 - 8 2033 62
AX763686.1 - 6 - 7 1872 62
BC027824.1 - 7 - 8 2003 62
BC035201.1 - 7 - 8 2015 62
BC108411.1 - 7 - 8 2067 62

View Full Product Details