The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019764.2 | NP_062738.2 | 9 - 10 | 2422 | 97 |
GenBank mRNA | AF175967.1 | - | 9 - 10 | 1705 | 97 |
AF175968.1 | - | 7 - 8 | 2562 | 97 | |
AK040912.1 | - | 9 - 10 | 2503 | 97 | |
AK077535.1 | - | 9 - 10 | 2418 | 97 | |
AK141348.1 | - | 9 - 10 | 2422 | 97 | |
AK164279.1 | - | 9 - 10 | 2491 | 97 | |
AK164819.1 | - | 10 - 11 | 2522 | 97 | |
AK169419.1 | - | 9 - 10 | 2422 | 97 | |
AX763686.1 | - | 8 - 9 | 2261 | 97 | |
BC027824.1 | - | 9 - 10 | 2392 | 97 | |
BC035201.1 | - | 9 - 10 | 2404 | 97 | |
BC108411.1 | - | 9 - 10 | 2456 | 97 |