The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019990.4 | NP_064374.1 | 6 - 7 | 779 | 75 |
GenBank mRNA | AB031550.1 | - | 7 - 8 | 956 | 75 |
BC046335.1 | - | 6 - 7 | 868 | 75 | |
BC058773.1 | - | 6 - 7 | 911 | 75 | |
BC080808.1 | - | 6 - 7 | 974 | 75 | |
BC094007.1 | - | 6 - 7 | 792 | 75 | |
BC116984.1 | - | 5 - 6 | 687 | 75 | |
BC120641.1 | - | 5 - 6 | 687 | 75 |