Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prg4
Entrez Gene ID
96875
Gene Name
proteoglycan 4 (megakaryocyte stimulating factor, articular superficial zone protein)
Gene Aliases
CACP, DOL54, JCAP, MSF, SZP, lubricin
Chromosome Location
Chr.1: 150449412 - 150466221 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110146.1 NP_001103616.1 7 - 8 591 75
NM_021400.3 NP_067375.3 8 - 9 2985 75
GenBank mRNA AB034730.1 - 9 - 10 2484 75
AK132597.1 - 2 - 3 731 75
AK149469.1 - 7 - 8 592 75
BC130021.1 - 6 - 7 555 75

View Full Product Details