The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026166.2 | NP_080442.2 | 2 - 3 | 378 | 72 |
GenBank mRNA | AK004670.1 | - | 2 - 3 | 336 | 72 |
BC055834.1 | - | 2 - 3 | 329 | 72 | |
CF171082.1 | - | 2 - 3 | 360 | 72 |