Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cep57l1
Entrez Gene ID
103268
Gene Name
centrosomal protein 57-like 1
Gene Aliases
2410017P07Rik, 4930484D11Rik, AI607327, cep57R
Chromosome Location
Chr.10: 41718840 - 41809868 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243075.1 NP_001230004.1 5 - 6 511 89
NM_029132.2 NP_083408.1 7 - 8 806 89
GenBank mRNA AF397908.1 - 7 - 8 734 89
AK006744.1 - 1 - 2 37 89
AK010530.1 - 7 - 8 757 89
AK012710.1 - 5 - 6 457 89
AK015619.1 - 6 - 7 721 89

View Full Product Details