The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026784.3 | NP_081060.2 | 1 - 2 | 347 | 73 |
GenBank mRNA | AK003607.1 | - | 1 - 2 | 150 | 73 |
AK150483.1 | - | 1 - 2 | 266 | 73 | |
AK168358.1 | - | 1 - 2 | 242 | 73 | |
BY266316.1 | - | 1 - 2 | 349 | 73 |