Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srpx2
Entrez Gene ID
68792
Gene Name
sushi-repeat-containing protein, X-linked 2
Gene Aliases
1110039C07Rik, SRP, SRPUL
Chromosome Location
Chr.X: 133908416 - 133932448 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026838.4 NP_081114.2 2 - 3 409 67
GenBank mRNA BC028307.1 - 2 - 3 138 67
CN539245.1 - 2 - 3 172 67

View Full Product Details