Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc46a3
Entrez Gene ID
71706
Gene Name
solute carrier family 46, member 3
Gene Aliases
1200006F02Rik
Chromosome Location
Chr.5: 147877846 - 147894864 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK004609.1 - 1 - 2 107 75
BC006902.1 - 1 - 2 103 75

View Full Product Details