The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027878.2 | NP_082154.2 | 3 - 4 | 496 | 68 |
GenBank mRNA | AK004552.1 | - | 3 - 4 | 496 | 68 |
AK151184.1 | - | 3 - 4 | 422 | 68 | |
BC021433.1 | - | 3 - 4 | 462 | 68 |