Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ints12
Entrez Gene ID
71793
Gene Name
integrator complex subunit 12
Gene Aliases
1110020M19Rik, 2810027J24Rik, 4930529N21Rik, A230056J18Rik, AI666778, Phf22
Chromosome Location
Chr.3: 133091953 - 133110988 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027927.3 NP_082203.1 3 - 4 348 123
GenBank mRNA AK003874.1 - 3 - 4 360 123
AK012829.1 - 3 - 4 361 123
AK155162.1 - 3 - 4 360 123
BC023801.1 - 3 - 4 348 123
BC046996.1 - 3 - 4 342 123

View Full Product Details