Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Nt5c2
- Entrez Gene ID
-
76952
- Gene Name
- 5'-nucleotidase, cytosolic II
- Gene Aliases
-
2010002I23Rik, CnII, ENSMUSG00000025039, Gm9751, Gmp, Nt5b, Pnt5, cN-II
- Chromosome Location
-
Chr.19: 46885390 - 47015189 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BC006028.1 |
- |
|
|
|
Chr. - - [-] on Build |
103 |
View Full Product Details