The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053110.4 | NP_444340.3 | 1 - 2 | 273 | 104 |
GenBank mRNA | AF322054.1 | - | 1 - 2 | 111 | 104 |
AJ251685.1 | - | 1 - 2 | 158 | 104 | |
AK044764.1 | - | 1 - 2 | 167 | 104 | |
AK076347.1 | - | 1 - 2 | 142 | 104 | |
AK079220.1 | - | - | 115 | 104 | |
AK149700.1 | - | 1 - 2 | 142 | 104 | |
AK149740.1 | - | 1 - 2 | 180 | 104 | |
AK150739.1 | - | 1 - 2 | 142 | 104 | |
AK154617.1 | - | 1 - 2 | 177 | 104 | |
AK159165.1 | - | 1 - 2 | 172 | 104 | |
AK159270.1 | - | 1 - 2 | 180 | 104 | |
AK159747.1 | - | 1 - 2 | 142 | 104 | |
AK159908.1 | - | 1 - 2 | 148 | 104 | |
AK170506.1 | - | 1 - 2 | 172 | 104 | |
AK171347.1 | - | 1 - 2 | 148 | 104 | |
AK171441.1 | - | 1 - 2 | 142 | 104 | |
AK171465.1 | - | 1 - 2 | 142 | 104 | |
AK171474.1 | - | 1 - 2 | 142 | 104 | |
AK171517.1 | - | 1 - 2 | 260 | 104 | |
AK171608.1 | - | 1 - 2 | 142 | 104 | |
AK171618.1 | - | 1 - 2 | 142 | 104 | |
BC026375.1 | - | 1 - 2 | 147 | 104 | |
BY205729.1 | - | 1 - 2 | 273 | 104 |