Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hrg
Entrez Gene ID
94175
Gene Name
histidine-rich glycoprotein
Gene Aliases
AI265597, AW413091, D16jh2, D18020, Hprg, Hrgp
Chromosome Location
Chr.16: 22951072 - 22961659 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053176.2 NP_444406.2 3 - 4 467 80
GenBank mRNA AB055897.1 - 3 - 4 425 80
AF194028.1 - 3 - 4 407 80
AY135662.1 - 3 - 4 428 80
BC011168.1 - 3 - 4 413 80
CB593922.1 - 3 - 4 439 80

View Full Product Details