Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp4f13
Entrez Gene ID
170716
Gene Name
cytochrome P450, family 4, subfamily f, polypeptide 13
Gene Aliases
0610030I10Rik, Cypf13
Chromosome Location
Chr.17: 32924688 - 32947415 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_130882.1 NP_570952.1 4 - 5 445 79
GenBank mRNA AF233643.1 - 4 - 5 445 79
BC003954.1 - 4 - 5 454 79

View Full Product Details