The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159581.1 | NP_001153053.1 | 3 - 4 | 246 | 101 |
NM_133250.2 | NP_573513.2 | 2 - 3 | 153 | 101 | |
GenBank mRNA | AB117937.1 | - | 1 - 2 | 115 | 101 |
AB117938.1 | - | 1 - 2 | 115 | 101 | |
AB117939.1 | - | 3 - 4 | 230 | 101 | |
AK017730.1 | - | 2 - 3 | 141 | 101 | |
AK077546.1 | - | 2 - 3 | 150 | 101 | |
AK156790.1 | - | 3 - 4 | 236 | 101 | |
AY007717.1 | - | 1 - 2 | 109 | 101 | |
BC057942.1 | - | 2 - 3 | 134 | 101 | |
CX213088.1 | - | 2 - 3 | 188 | 101 |