Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Erg
Entrez Gene ID
13876
Gene Name
avian erythroblastosis virus E-26 (v-ets) oncogene related
Gene Aliases
D030036I24Rik
Chromosome Location
Chr.16: 95359169 - 95586593 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB073078.1 - 2 - 3 186 71
AB073079.1 - 2 - 3 187 71
AB073080.1 - 2 - 3 186 71
AK050922.1 - 3 - 4 287 71
BC145176.1 - 1 - 2 77 71
BC145850.1 - 1 - 2 77 71

View Full Product Details