Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hiatl1
Entrez Gene ID
66631
Gene Name
hippocampus abundant transcript-like 1
Gene Aliases
5730414C17Rik, AU045608, AW539692
Chromosome Location
Chr.13: 65065030 - 65113000 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083901.1 NP_001077370.1 10 - 11 1304 65
NM_133680.3 NP_598441.3 9 - 10 1401 65
GenBank mRNA AK146596.1 - 10 - 11 1297 65
BC018395.1 - 2 - 3 189 65
BC033469.1 - 9 - 10 1363 65

View Full Product Details