The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159632.1 | NP_001153104.1 | 7 - 8 | 646 | 62 |
NM_133699.2 | NP_598460.1 | 6 - 7 | 616 | 62 | |
GenBank mRNA | AB088357.1 | - | 6 - 7 | 601 | 62 |
AK004157.1 | - | 3 - 4 | 331 | 62 | |
BC056636.1 | - | 7 - 8 | 619 | 62 | |
BC064071.1 | - | 6 - 7 | 585 | 62 |