Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kptn
Entrez Gene ID
70394
Gene Name
kaptin
Gene Aliases
2310042D10Rik, 2E4, C030013F01Rik
Chromosome Location
Chr.7: 16119876 - 16127516 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC018316.1 - 4 - 5 496 85

View Full Product Details