Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccdc71
Entrez Gene ID
72454
Gene Name
coiled-coil domain containing 71
Gene Aliases
2600016J21Rik
Chromosome Location
Chr.9: 108452571 - 108465945 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133744.4 NP_598505.1 1 - 2 105 64
GenBank mRNA AK011229.1 - 1 - 2 71 64
AK044601.1 - 1 - 2 71 64
AK079333.1 - 1 - 2 78 64
AK086560.1 - 1 - 2 71 64
AK136982.1 - 1 - 2 58 64
AK149878.1 - 1 - 2 71 64
BC018518.1 - 1 - 2 56 64
CJ146452.1 - 1 - 2 105 64

View Full Product Details