The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133746.5 | NP_598507.3 | 2 - 3 | 1106 | 96 |
GenBank mRNA | AK012923.1 | - | 2 - 3 | 1031 | 96 |
AK036961.1 | - | - | 1082 | 96 | |
AK040739.1 | - | 2 - 3 | 1107 | 96 | |
AK089278.1 | - | 2 - 3 | 1107 | 96 | |
AK170068.1 | - | 2 - 3 | 1077 | 96 | |
BC019197.1 | - | 2 - 3 | 1028 | 96 |