The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133803.2 | NP_598564.2 | 17 - 18 | 2114 | 72 |
GenBank mRNA | AK031020.1 | - | 17 - 18 | 2079 | 72 |
AK087994.1 | - | 17 - 18 | 2078 | 72 | |
AK150101.1 | - | 17 - 18 | 2074 | 72 | |
AK150126.1 | - | 17 - 18 | 2068 | 72 | |
AK166842.1 | - | 17 - 18 | 2079 | 72 | |
AK168195.1 | - | 17 - 18 | 2067 | 72 | |
AK200466.1 | - | 1 - 2 | 143 | 72 | |
BC004600.1 | - | 16 - 17 | 2056 | 72 | |
BC066817.1 | - | 17 - 18 | 2065 | 72 |