Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Zbtb48
Entrez Gene ID
100090
Gene Name
zinc finger and BTB domain containing 48
Gene Aliases
0610011D15Rik, AI327031, Hkr3
Chromosome Location
Chr.4: 152019773 - 152027671 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133879.2 NP_598640.2 9 - 10 1791 63
GenBank mRNA AK027912.1 - 8 - 9 2275 63
AM161454.1 - 9 - 10 1739 63
BC005515.1 - 8 - 9 1896 63

View Full Product Details