The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113362.1 | NP_001106833.1 | 12 - 13 | 1889 | 148 |
NM_001113364.1 | NP_001106835.1 | 9 - 10 | 1003 | 148 | |
NM_133910.3 | NP_598671.3 | 11 - 12 | 1708 | 148 | |
GenBank mRNA | AB064521.1 | - | 9 - 10 | 866 | 148 |
AB093293.1 | - | 10 - 11 | 1502 | 148 | |
AK013092.1 | - | 9 - 10 | 929 | 148 | |
AK029986.1 | - | 12 - 13 | 1889 | 148 | |
BC011420.1 | - | 9 - 10 | 881 | 148 | |
BC042515.1 | - | 11 - 12 | 1684 | 148 | |
BC096446.1 | - | 11 - 12 | 1700 | 148 |