Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfxl1
Entrez Gene ID
100978
Gene Name
nuclear transcription factor, X-box binding-like 1
Gene Aliases
1700012H24Rik, AW538212, D430033A06Rik, DNABF, GCF, Gm1058, TCF9
Chromosome Location
Chr.5: 72513301 - 72559707 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133921.2 NP_598682.2 21 - 22 2584 61
GenBank mRNA AK132320.1 - 20 - 21 2494 61
BC022652.1 - 5 - 6 584 61
BC032981.1 - 1 - 2 267 61

View Full Product Details