The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133927.1 | NP_598688.1 | 1 - 2 | 196 | 57 |
GenBank mRNA | AK142721.1 | - | 1 - 2 | 216 | 57 |
AK147728.1 | - | 1 - 2 | 198 | 57 | |
AK159637.1 | - | 1 - 2 | 205 | 57 | |
BC014833.1 | - | 1 - 2 | 196 | 57 | |
BC025492.1 | - | 1 - 2 | 216 | 57 |