Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fig4
Entrez Gene ID
103199
Gene Name
FIG4 homolog (S. cerevisiae)
Gene Aliases
A530089I17Rik, AI326867, Sac3
Chromosome Location
Chr.10: 41188172 - 41303241 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133999.1 NP_598760.1 12 - 13 1581 64
GenBank mRNA AK041196.1 - 12 - 13 1582 64
AK172130.1 - 12 - 13 1575 64
AK172341.1 - 12 - 13 1546 64
AK172926.1 - 5 - 6 629 64
AK181110.1 - 1 - 2 45 64
AK204518.1 - 1 - 2 109 64
BC015295.1 - 12 - 13 1581 64
BC031887.1 - 12 - 13 1580 64

View Full Product Details