The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134066.2 | NP_598827.1 | 7 - 8 | 884 | 126 |
GenBank mRNA | AB059565.1 | - | 7 - 8 | 884 | 126 |
AK153898.1 | - | 7 - 8 | 902 | 126 | |
BC034259.1 | - | 6 - 7 | 803 | 126 |