Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Akr1c18
Entrez Gene ID
105349
Gene Name
aldo-keto reductase family 1, member C18
Gene Aliases
AW146047
Chromosome Location
Chr.13: 4132615 - 4150651 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134066.2 NP_598827.1 8 - 9 961 131
GenBank mRNA AB059565.1 - 8 - 9 961 131
AK153898.1 - 8 - 9 979 131
BC034259.1 - 7 - 8 880 131

View Full Product Details