Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ankrd32
Entrez Gene ID
105377
Gene Name
ankyrin repeat domain 32
Gene Aliases
2700017A04Rik, AW545819, Brctd1, Brctx, C730024G01Rik
Chromosome Location
Chr.13: 77043088 - 77135468 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_134071.3 NP_598832.3 19 - 20 2620 82
GenBank mRNA AB221577.1 - 5 - 6 605 82
BC009101.1 - 4 - 5 463 82
BC024900.1 - 7 - 8 905 82
BC083095.1 - 9 - 10 1232 82

View Full Product Details