Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rcbtb2
Entrez Gene ID
105670
Gene Name
regulator of chromosome condensation (RCC1) and BTB (POZ) domain containing protein 2
Gene Aliases
2610028E02Rik, 2810420M18Rik, AI448345, AW240694, Chc1l, RC/BTB2
Chromosome Location
Chr.14: 73138773 - 73184055 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170694.1 NP_001164165.1 9 - 10 1068 97
NM_134083.4 NP_598844.3 9 - 10 1063 97
NR_033185.1 10 - 11 1448 97
GenBank mRNA AK013540.1 - - 1173 97
AK031242.1 - 9 - 10 1029 97
AK031248.1 - 9 - 10 1034 97
AK147784.1 - 10 - 11 1069 97
AK160881.1 - 10 - 11 1255 97
BC003224.1 - 7 - 8 861 97
JN086338.1 - 8 - 9 984 97
JN086339.1 - 9 - 10 1222 97

View Full Product Details