The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134093.2 | NP_598854.1 | 7 - 8 | 1004 | 67 |
GenBank mRNA | AF287293.1 | - | 7 - 8 | 944 | 67 |
AF401483.1 | - | 7 - 8 | 837 | 67 | |
AK045256.1 | - | 7 - 8 | 824 | 67 | |
AK046496.1 | - | 5 - 6 | 904 | 67 | |
AK080221.1 | - | 7 - 8 | 924 | 67 | |
AK158718.1 | - | 6 - 7 | 781 | 67 | |
AK169341.1 | - | 7 - 8 | 924 | 67 | |
AK190806.1 | - | 1 - 2 | 92 | 67 | |
BC021361.1 | - | 4 - 5 | 676 | 67 |